4-Bromoveratrole 98%

Supplier: Ambeed
Warning

A139693-100G A139693-10G A139693-25G A139693-500G A139693-5G
77109-758EA 46.35 USD
77109-758 77109-760 77109-762 77109-764 77109-766
4-Bromoveratrole 98%
4-Bromoveratrole

Precaución: for Research Use Only

Formula: C₈H₉BrO₂
MDL Number: MFCD00008381
CAS Number: 2859-78-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR