3-Bromo-5-nitrosalicylaldehyde 98%

Supplier: Ambeed
Danger

A139047-100G A139047-10G A139047-1G A139047-25G A139047-5G
77109-722EA 303.85 USD
77109-722 77109-724 77109-726 77109-728 77109-730
3-Bromo-5-nitrosalicylaldehyde 98%
3-Bromo-5-nitrosalicylaldehyde

Precaución: for Research Use Only

Formula: C₇H₄BrNO₄
MDL Number: MFCD00051833
CAS Number: 16789-84-7
UN: 2811
ADR: 9,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR