RO-3306 ≥98%

Supplier: Ambeed
Warning

A138869-10MG A138869-1MG A138869-5MG
77109-714EA 20.6 USD
77109-714 77109-716 77109-718
RO-3306 ≥98%
RO-3306

Precaución: for Research Use Only

Formula: C₁₈H₁₃N₃OS₂
MDL Number: MFCD17392573
CAS Number: 872573-93-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR