3-Amino-2-chloro-5-fluoropyridine 98%
Supplier: Ambeed
|
Warning
|
A138340-100MG
A138340-10G
A138340-1G
A138340-250MG
A138340-5G
77109-698EA
7.21
USD
77109-698
77109-700
77109-702
77109-704
77109-706
3-Amino-2-chloro-5-fluoropyridine 98%
3-Amino-2-chloro-5-fluoropyridine
Precaución: for Research Use Only
Formula:
C₅H₄ClFN₂ |
MDL Number:
MFCD08277275 CAS Number: 884495-37-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...