1-Butylpyrazole 98%

Supplier: Ambeed
Warning

A138057-1G A138057-250MG A138057-5G
77109-684EA 56.65 USD
77109-684 77109-686 77109-688
1-Butylpyrazole 98%
1-Butylpyrazole

Precaución: for Research Use Only

Formula: C₇H₁₂N₂
MDL Number: MFCD08701137
CAS Number: 52096-24-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR