9-Phenylacridine 98%

Supplier: Ambeed
Warning

A137990-10G A137990-1G A137990-25G A137990-5G
77109-676EA 25.75 USD
77109-676 77109-678 77109-680 77109-682
9-Phenylacridine 98%
9-Phenylacridine

Precaución: for Research Use Only

Formula: C₁₉H₁₃N
MDL Number: MFCD00092562
CAS Number: 602-56-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR