4-Amino-6-methylpyrimidine-5-carbonitrile 95%

Supplier: Ambeed
Warning

A137448-250MG A137448-5G
77109-608EA 93.36 USD
77109-608 77109-610
4-Amino-6-methylpyrimidine-5-carbonitrile 95%
4-Amino-6-methylpyrimidine-5-carbonitrile

Precaución: for Research Use Only

Formula: C₆H₆N₄
MDL Number: MFCD01312248
CAS Number: 76574-44-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR