2-Amino-5-nitro-4-picoline 98%

Supplier: Ambeed
Warning

Synonyms: 4-Methyl-5-nitro-2-pyridylamine, 4-Methyl-5-nitropyridin-2-amine, 2-Amino-4-methyl-5-nitropyridine

A137410-100G A137410-25G A137410-500G A137410-5G
77109-606EA 72.15 USD
77109-606 77611-248 77611-250 77611-252
2-Amino-5-nitro-4-picoline 98%
2-Amino-5-nitro-4-picoline

Precaución: for Research Use Only

Formula: C₆H₇N₃O₂
MW: 153.14 g/mol
MDL Number: MFCD00010692
CAS Number: 21901-40-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR