Alendronate sodium trihydrate 98%
Supplier: Ambeed
|
Warning
|
A136794-10G
A136794-25G
A136794-5G
A136794-100G
77109-588EA
11.67
USD
77109-588
77109-590
77109-592
77615-086
Alendronate sodium trihydrate 98%
Alendronate sodium trihydrate
Precaución: for Research Use Only
Formula:
C₄H₁₂NaNO₇P₂·3H₂O |
MDL Number:
MFCD01748233 CAS Number: 121268-17-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...