3'-Aminoacetophenone 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
m-Aminoacetophenone, 1-(3-Aminophenyl)ethanone
A136182-1000G
A136182-100G
A136182-10G
A136182-25G
A136182-500G
77109-548EA
223.85
USD
77109-548
77109-550
77109-552
77109-554
77109-556
3'-Aminoacetophenone 98%
3'-Aminoacetophenone
Precaución: for Research Use Only
Formula:
C₈H₉NO MW: 135.17 g/mol |
MDL Number:
MFCD00007796 CAS Number: 99-03-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...