4-Pyridone 97%

Supplier: Ambeed
Warning

Synonyms: 4-Hydroxypyridine, γ-Pyridone, 4-Pyridinol, Pyridin-4-ol, 1,4-Dihydro-4-Pyridinone

A136051-1000G A136051-100G A136051-10G A136051-500G A136051-5G A136051-25G
77109-538EA 270.88 USD
77109-538 77109-540 77109-542 77109-544 77109-546 77315-148
4-Pyridone 97%
4-Pyridone

Precaución: for Research Use Only

Formula: C₅H₅NO
MW: 95.1 g/mol
Boiling Pt: 381
MDL Number: MFCD00006419
CAS Number: 626-64-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR