HMN-214 98%

Supplier: Ambeed
Warning

Synonyms: HMN214, IVX214, (E)-4-(2-(N-((4-methoxyphenyl)sulphonyl)acetamido)styryl)pyridine 1-oxide, N-(4-methoxyphenyl)sulphonyl-N-[2-[(E)-2-(1-oxidopyridin-1-ium-4-yl)ethenyl]phenyl]acetamide, IVX 214, HMN-214, HMN 214, IVX-214

A136004-10MG A136004-1MG A136004-25MG A136004-50MG A136004-5MG A136004-100MG A136004-250MG
77109-528EA 78.51 USD
77109-528 77109-530 77109-532 77109-534 77109-536 77685-174 77685-175
HMN-214 98%
HMN-214

Precaución: for Research Use Only

Formula: C₂₂H₂₀N₂O₅S
MW: 424.47 g/mol
MDL Number: MFCD12756255
CAS Number: 173529-46-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR