P5091 99+%

Supplier: Ambeed
Danger

Synonyms: 1-(5-((2,3-Dichlorophenyl)thio)-4-nitrothiophen-2-yl)ethan-1-one, P005091

A135201-100MG A135201-10MG A135201-1MG A135201-50MG A135201-5MG
77109-476EA 78.51 USD
77109-476 77109-478 77109-480 77109-482 77109-484
P5091 99+%
P5091

Precaución: for Research Use Only

Formula: C₁₂H₇Cl₂NO₃S₂
MW: 348.22 g/mol
MDL Number: MFCD00123202
CAS Number: 882257-11-6
UN: 2811
ADR: 6.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR