STF-62247 95%

Supplier: Ambeed
Warning

Synonyms: 4-(Pyridin-4-yl)-N-(m-tolyl)thiazol-2-amine, N-(3-Methylphenyl)-4-(4-pyridinyl)-2-thiazolamine

A134944-10MG A134944-1MG A134944-25MG A134944-50MG
77109-460EA 23.34 USD
77109-460 77109-462 77109-464 77109-466
STF-62247 95%
STF-62247

Precaución: for Research Use Only

Formula: C₁₅H₁₃N₃S
MW: 267.35 g/mol
MDL Number: MFCD01121974
CAS Number: 315702-99-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR