3,9-Di(cyclohex-3-en-1-yl)-2,4,8,10-tetraoxaspiro[5.5]undecane 95%

Supplier: Ambeed
Warning

A134163-10G A134163-25G
77109-422EA 6.37 USD
77109-422 77109-424
3,9-Di(cyclohex-3-en-1-yl)-2,4,8,10-tetraoxaspiro[5.5]undecane 95%
3,9-Di(cyclohex-3-en-1-yl)-2,4,8,10-tetraoxaspiro[5.5]undecane

Precaución: for Research Use Only

Formula: C₁₉H₂₈O₄
MW: 320.42 g/mol
MDL Number: MFCD00023815
CAS Number: 6600-31-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR