(S)-1-Methyl-3-pyrrolidinol 97%

Supplier: Ambeed
Warning

Synonyms: (S)-3-Hydroxy-1-methylpyrrolidine, (S)-(+)-1-Methyl-3-pyrrolidinol

A133564-100G A133564-10G A133564-1G A133564-250MG A133564-25G A133564-5G
77109-384EA 519.85 USD
77109-384 77109-386 77109-388 77109-390 77109-392 77109-394
(S)-1-Methyl-3-pyrrolidinol 97%
(S)-1-Methyl-3-pyrrolidinol

Precaución: for Research Use Only

Formula: C₅H₁₁NO
MW: 101.15 g/mol
MDL Number: MFCD03788748
CAS Number: 104641-59-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR