2-Chloro-5-formyl-4-picoline 97%

Supplier: Ambeed
Warning

A133404-1G A133404-250MG A133404-5G
77109-378EA 23.68 USD
77109-378 77109-380 77109-382
2-Chloro-5-formyl-4-picoline 97%
2-Chloro-5-formyl-4-picoline

Precaución: for Research Use Only

Formula: C₇H₆ClNO
MW: 155.58 g/mol
MDL Number: MFCD07368881
CAS Number: 884495-38-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR