(R)-3-Amino-3-(3-nitrophenyl)propionic acid 97%

Supplier: Ambeed
Warning

A133185-1G A133185-250MG A133185-5G
77109-370EA 97.61 USD
77109-370 77109-372 77109-374
(R)-3-Amino-3-(3-nitrophenyl)propionic acid 97%
(R)-3-Amino-3-(3-nitrophenyl)propionic acid

Precaución: for Research Use Only

Formula: C₉H₁₀N₂O₄
MW: 210.18 g/mol
MDL Number: MFCD04113660
CAS Number: 787544-61-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR