(5-Fluorobiphenyl-2-yl)methanamine 95%

Supplier: Ambeed
Danger

A133002-1G A133002-5G
77109-356EA 89.61 USD
77109-356 77109-358
(5-Fluorobiphenyl-2-yl)methanamine 95%
(5-Fluorobiphenyl-2-yl)methanamine

Precaución: for Research Use Only

Formula: C₁₃H₁₂FN
MDL Number: MFCD14701931
CAS Number: 1002557-11-0
UN: 2811
ADR: 6.1,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR