(E)-2-(2-Nitroethenyl)thiophene 98%

Supplier: Ambeed
Danger

A133000-1G A133000-25G A133000-5G
77109-350EA 23.34 USD
77109-350 77109-352 77109-354
(E)-2-(2-Nitroethenyl)thiophene 98%
(E)-2-(2-Nitroethenyl)thiophene

Precaución: for Research Use Only

Formula: C₆H₅NO₂S
MW: 155.17 g/mol
MDL Number: MFCD00069434
CAS Number: 34312-77-1
UN: 2811
ADR: 6.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR