Calcitriol 98%

Supplier: Ambeed
Danger

Synonyms: 1,25-Dihydroxycholecalciferol, (1R,3S,Z)-5-(2-((1R,3aS,7aR,E)-1-((R)-6-Hydroxy-6-methylheptan-2-yl)-7a-methyloctahydro-4H-inden-4-ylidene)ethylidene)-4-methylenecyclohexane-1,3-diol, 1alpha,25-Dihydroxyvitamin D3

A132740-100MG A132740-50MG A132740-1G A132740-10MG A132740-5MG
77109-346EA 479.53 USD
77109-346 77109-348 77566-586 77615-056 77685-150
Calcitriol 98%
Calcitriol

Precaución: For research use only.

Formula: C₂₇H₄₄O₃
MW: 416.646 g/mol
MDL Number: MFCD00867079
CAS Number: 32222-06-3
UN: 2811
ADR: 6.1,Ⅰ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR