1-Ethynyl-3-nitrobenzene 98%

Supplier: Ambeed
Warning

Synonyms: 3-Nitrophenylacetylene, 1-Ethynyl-3-nitrobenzene, (3-Nitrophenyl)ethyne, (3-Nitrophenyl)acetylene

A132226-100G A132226-10G A132226-1G A132226-250MG A132226-25G A132226-5G
77109-326EA 176.14 USD
77109-326 77109-328 77109-330 77109-332 77109-334 77109-336
1-Ethynyl-3-nitrobenzene 98%
1-Ethynyl-3-nitrobenzene

Precaución: for Research Use Only

Formula: C₈H₅NO₂
MDL Number: MFCD02181150
CAS Number: 3034-94-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR