Imidazo[1,2-a]pyridine-3-carbaldehyde 98%

Supplier: Ambeed
Warning

A131677-100MG A131677-1G A131677-25G A131677-5G A131677-250MG
77109-274EA 8.23 USD
77109-274 77109-276 77109-278 77109-280 77685-145
Imidazo[1,2-a]pyridine-3-carbaldehyde 98%
Imidazo[1,2-a]pyridine-3-carbaldehyde

Precaución: for Research Use Only

Formula: C₈H₆N₂O
MDL Number: MFCD07778354
CAS Number: 6188-43-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR