5-Bromo-m-xylene 98%

Supplier: Ambeed
Warning

A131255-1000G A131255-100G A131255-10G A131255-500G A131255-5G A131255-25G
77109-258EA 160.68 USD
77109-258 77109-260 77109-262 77109-264 77109-266 77314-034
5-Bromo-m-xylene 98%
5-Bromo-m-xylene

Precaución: for Research Use Only

Formula: C₈H₉Br
MDL Number: MFCD00000087
CAS Number: 556-96-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR