2,3,6-Trifluorobenzoic acid 98%

Supplier: Ambeed
Warning

A130985-10G A130985-1G A130985-25G A130985-5G
77109-248EA 25.75 USD
77109-248 77109-250 77109-252 77109-254
2,3,6-Trifluorobenzoic acid 98%
2,3,6-Trifluorobenzoic acid

Precaución: for Research Use Only

Formula: C₇H₃F₃O₂
MDL Number: MFCD00002409
CAS Number: 2358-29-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR