Ozagrel 98%

Supplier: Ambeed
Warning

A130858-100MG A130858-10MG A130858-1G A130858-1MG A130858-250MG A130858-50MG A130858-5G A130858-5MG
77109-230EA 20.6 USD
77109-230 77109-232 77109-234 77109-236 77109-238 77109-240 77109-242 77109-244
Ozagrel 98%
Ozagrel

Precaución: for Research Use Only

Formula: C₁₃H₁₂N₂O₂
MDL Number: MFCD00868231
CAS Number: 82571-53-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR