4,5-Difluoro-2-hydroxybenzoic acid 95%

Supplier: Ambeed
Warning

A130371-100MG A130371-250MG
77109-206EA 49.44 USD
77109-206 77109-208
4,5-Difluoro-2-hydroxybenzoic acid 95%
4,5-Difluoro-2-hydroxybenzoic acid

Precaución: for Research Use Only

Formula: C₇H₄F₂O₃
MDL Number: MFCD13185554
CAS Number: 205533-31-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR