L(+)-Cysteine hydrochloride monohydrate 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
(R)-(+)-Cysteine hydrochloride monohydrate, (R)-2-Amino-3-mercaptopropanoic acid hydrochloride hydrate, L-Cysteine HCI Monohydrate, H-Cys-OH.HCl.H2O, Cysteine Hydrochloride, Monohydrate, Cysteine Hydrochloride monohydrate
A130206-100G
A130206-1000G
A130206-500G
77109-196EA
12.36
USD
77109-196
77109-198
77611-152
77685-129
L(+)-Cysteine hydrochloride monohydrate 98%
L(+)-Cysteine hydrochloride monohydrate
Precaución: for Research Use Only
Formula:
HSCH₂CH(NH₂)COOH·HCl·H₂O MW: 175.64 g/mol |
MDL Number:
MFCD00065606 CAS Number: 7048-04-6 Merck Index: 13,02810 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...