3-Fluoro-4-nitrobenzaldehyde 95%

Supplier: Ambeed
Warning

A130091-1G A130091-5G A130091-100G
77109-194EA 12.36 USD
77109-194 77566-530 77685-126
3-Fluoro-4-nitrobenzaldehyde 95%
3-Fluoro-4-nitrobenzaldehyde

Precaución: For research use only.

Formula: C₇H₄NO₃F
MDL Number: MFCD00968940
CAS Number: 160538-51-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR