tert-Butyl triphenylphosphoranylidenecarbamate 98%

Supplier: Ambeed
Warning

Synonyms: N-Boc-imino(triphenyl)phosphorane, (Triphenylphosphoranylidene)carbamic acid tert-Butyl ester, N-Boc-Imino-(triphenyl)phosphorane, tert-Butyl (Triphenylphosphoranylidene)carbamate, N-(tert-Butoxycarbonyl)imino(triphenyl)phosphorane

A130043-100G A130043-10G A130043-1G A130043-25G A130043-500G A130043-5G
77109-180EA 69.01 USD
77109-180 77109-182 77109-184 77109-186 77109-188 77109-190
tert-Butyl triphenylphosphoranylidenecarbamate 98%
tert-Butyl triphenylphosphoranylidenecarbamate

Precaución: for Research Use Only

Formula: C₂₃H₂₄NO₂P
MW: 377.43 g/mol
MDL Number: MFCD00798174
CAS Number: 68014-21-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR