2-Ethyl-3-methylpyrazine 98%

Supplier: Ambeed
Danger

A130011-10G A130011-1G A130011-5G
77109-174EA 8.49 USD
77109-174 77109-176 77109-178
2-Ethyl-3-methylpyrazine 98%
2-Ethyl-3-methylpyrazine

Precaución: for Research Use Only

Formula: C₇H₁₀N₂
MW: 122.17 g/mol
MDL Number: MFCD00006150
CAS Number: 15707-23-0
UN: 1993
ADR: 3,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR