2-Fluoro-4-(hydroxymethyl)benzoic acid 98%

Supplier: Ambeed
Warning

Synonyms: (4-Carboxy-3-fluorophenyl)methanol, 4-Carboxy-3-fluorobenzyl alcohol

A129983-100MG A129983-250MG
77109-170EA 31.83 USD
77109-170 77109-172
2-Fluoro-4-(hydroxymethyl)benzoic acid 98%
2-Fluoro-4-(hydroxymethyl)benzoic acid

Precaución: for Research Use Only

Formula: C₈H₇FO₃
MW: 170.138 g/mol
MDL Number: MFCD08741375
CAS Number: 214554-18-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR