2-Fluoro-3-nitrobenzaldehyde 95%

Supplier: Ambeed
Warning

A129711-100MG A129711-250MG A129711-1G A129711-5G
77109-154EA 10.31 USD
77109-154 77109-156 77685-124 77685-125
2-Fluoro-3-nitrobenzaldehyde 95%
2-Fluoro-3-nitrobenzaldehyde

Precaución: for Research Use Only

Formula: C₇H₄FNO₃
MDL Number: MFCD08669885
CAS Number: 96516-29-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR