(S)-2-Boc-aminomethylpyrrolidine 95%

Supplier: Ambeed
Warning

Synonyms: (S)-tert-butyl pyrrolidin-2-ylmethylcarbamate, (S)-tert-Butyl (pyrrolidin-2-ylmethyl)carbamate

A129606-250MG A129606-5G
77109-146EA 27.59 USD
77109-146 77611-148
(S)-2-Boc-aminomethylpyrrolidine 95%
(S)-2-Boc-aminomethylpyrrolidine

Precaución: for Research Use Only

Formula: C₁₀H₂₀N₂O₂
MW: 200.2780 g/mol
MDL Number: MFCD06796572
CAS Number: 141774-70-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR