4-Hydroxy-2-(trifluoromethoxy)benzaldehyde 98%
Supplier: Ambeed
|
Warning
|
A129574-1G
A129574-250MG
A129574-5G
77109-140EA
46.68
USD
77109-140
77109-142
77109-144
4-Hydroxy-2-(trifluoromethoxy)benzaldehyde 98%
4-Hydroxy-2-(trifluoromethoxy)benzaldehyde
Precaución: for Research Use Only
Formula:
C₈H₅F₃O₃ MW: 206.11 g/mol |
MDL Number:
MFCD06797920 CAS Number: 1017083-37-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...