5-Bromo-2-chlorobenzonitrile 97%
Supplier: Ambeed
|
Warning
|
A129533-10G
A129533-25G
A129533-5G
77109-134EA
15.45
USD
77109-134
77109-136
77109-138
5-Bromo-2-chlorobenzonitrile 97%
5-Bromo-2-chlorobenzonitrile
Precaución: for Research Use Only
Formula:
C₇H₃BrClN |
MDL Number:
MFCD00672948 CAS Number: 57381-44-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...