3-Hydroxybenzonitrile 98%

Supplier: Ambeed
Warning

Synonyms: m-Hydroxybenzonitrile, m-Cyanophenol, 3-Cyanophenol

A129461-10G A129461-1G A129461-5G
77109-130EA 18.54 USD
77109-130 77109-132 77592-498
3-Hydroxybenzonitrile 98%
3-Hydroxybenzonitrile

Precaución: for Research Use Only

Formula: C₇H₅NO
MDL Number: MFCD00002252
CAS Number: 873-62-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR