Menadione (2-methyl-1,4-naphthoquinone, vitamin K3) 98%

Supplier: Ambeed
Warning

Synonyms: Menadione, 1,4-Dihydro-2-methylnaphthalene-1,4-dione, 2-Methylnaphthalene-1,4-dione

A129193-100G A129193-1G A129193-250MG A129193-25G A129193-500G
77109-114EA 57.29 USD
77109-114 77109-116 77109-118 77109-120 77109-122
Menadione (2-methyl-1,4-naphthoquinone, vitamin K3) 98%
Menadione (2-methyl-1,4-naphthoquinone, vitamin K3)

Precaución: for Research Use Only

Formula: C₁₁H₈O₂
MW: 172.19 g/mol
MDL Number: MFCD00001681
CAS Number: 58-27-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR