6-(Methoxycarbonyl)-1H-indazole-3-carboxylic acid 95%

Supplier: Ambeed
Warning

A128997-1G A128997-250MG A128997-5G
77109-096EA 405.27 USD
77109-096 77109-098 77109-100
6-(Methoxycarbonyl)-1H-indazole-3-carboxylic acid 95%
6-(Methoxycarbonyl)-1H-indazole-3-carboxylic acid

Precaución: for Research Use Only

Formula: C₁₀H₈N₂O₄
MW: 220.18 g/mol
MDL Number: MFCD07781536
CAS Number: 885522-60-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR