4-Methoxy-6-methyl-2-(methylthio)pyrimidine 95%

Supplier: Ambeed
Warning

A128973-100MG A128973-1G A128973-250MG
77109-090EA 9.55 USD
77109-090 77109-092 77109-094
4-Methoxy-6-methyl-2-(methylthio)pyrimidine 95%
4-Methoxy-6-methyl-2-(methylthio)pyrimidine

Precaución: for Research Use Only

Formula: C₇H₁₀N₂OS
MW: 170.23 g/mol
MDL Number: MFCD10574964
CAS Number: 55749-33-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR