Ethyl 2-(4-(hydroxymethyl)piperidin-1-yl)pyrimidine-5-carboxylate 95%
Supplier: Ambeed
|
Warning
|
A128879-100MG
A128879-1G
A128879-250MG
A128879-25G
77109-080EA
16.98
USD
77109-080
77109-082
77109-084
77109-086
Ethyl 2-(4-(hydroxymethyl)piperidin-1-yl)pyrimidine-5-carboxylate 95%
Ethyl 2-(4-(hydroxymethyl)piperidin-1-yl)pyrimidine-5-carboxylate
Precaución: for Research Use Only
Formula:
C₁₃H₁₉N₃O₃ MW: 265.31 g/mol |
MDL Number:
MFCD09991532 CAS Number: 875318-46-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...