2-Bromo-4-(trifluoromethyl)phenol 98%

Supplier: Ambeed
Warning

Synonyms: 3-Bromo-4-hydroxybenzotrifluoride

A128665-100G A128665-10G A128665-1G A128665-25G A128665-5G
77109-056EA 233.4 USD
77109-056 77109-058 77109-060 77109-062 77109-064
2-Bromo-4-(trifluoromethyl)phenol 98%
2-Bromo-4-(trifluoromethyl)phenol

Precaución: for Research Use Only

Formula: C₇H₄BrF₃O
MW: 241.01 g/mol
MDL Number: MFCD06657954
CAS Number: 81107-97-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR