1-(3-Bromophenyl)piperidine 96%

Supplier: Ambeed
Warning

A128260-10G A128260-250MG
77109-038EA 231.28 USD
77109-038 77109-040
1-(3-Bromophenyl)piperidine 96%
1-(3-Bromophenyl)piperidine

Precaución: for Research Use Only

Formula: C₁₁H₁₄BrN
MW: 240.1396 g/mol
MDL Number: MFCD04108532
CAS Number: 84964-24-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR