5-Bromo-1,2,3-trimethoxybenzene 98%

Supplier: Ambeed
Warning

Synonyms: 1-Bromo-3,4,5-trimethoxybenzene

A127797-100G A127797-1G A127797-25G A127797-5G
77109-012EA 144.2 USD
77109-012 77109-014 77109-016 77109-018
5-Bromo-1,2,3-trimethoxybenzene 98%
5-Bromo-1,2,3-trimethoxybenzene

Precaución: for Research Use Only

Formula: C₉H₁₁BrO₃
MDL Number: MFCD00017169
CAS Number: 2675-79-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR