Boc-L-Pro-OMe 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
Boc-Pro-OMe, N-Boc-L-proline methyl ester, N-(tert-Butoxycarbonyl)-L-proline methyll ester
A127658-100G
A127658-10G
A127658-500G
A127658-5G
A127658-25G
77109-004EA
56.64
USD
77109-004
77109-006
77109-008
77109-010
77313-250
Boc-L-Pro-OMe 97%
Boc-L-Pro-OMe
Precaución: for Research Use Only
Formula:
C₁₁H₁₉NO₄ |
MDL Number:
MFCD01075092 CAS Number: 59936-29-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...