2-Chloro-4,6-dimethoxypyrimidine 95%

Supplier: Ambeed
Warning

A127420-1000G A127420-100G A127420-10G A127420-25G A127420-500G A127420-5G
77108-974EA 563.41 USD
77108-974 77108-976 77108-978 77108-980 77108-982 77108-984 77108-986
2-Chloro-4,6-dimethoxypyrimidine 95%
2-Chloro-4,6-dimethoxypyrimidine

Precaución: for Research Use Only

Formula: C₆H₇ClN₂O₂
MDL Number: MFCD00274530
CAS Number: 13223-25-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR