D-Asparagine monohydrate 97%

Supplier: Ambeed
Warning

Synonyms: (R)-2-Aminosuccinamic acid hydrate, D-2-Aminosuccinamic acid, H-D-Asn-OH·H₂O, (R)-2,4-Diamino-4-oxobutanoic acid hydrate

A127408-25G A127408-5G
77108-970EA 14.86 USD
77108-970 77108-972
D-Asparagine monohydrate 97%
D-Asparagine monohydrate

Precaución: for Research Use Only

Formula: C₄H₁₀N₂O₄
MW: 150.14 g/mol (132.12 anhy)
MDL Number: MFCD00149558
CAS Number: 5794-24-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR