Boc-S-trityl-D-cysteine 97%

Supplier: Ambeed
Warning

Synonyms: N-Boc-S-trityl-D-cysteine, Boc-D-Cys(Trt)-OH

A126964-100G A126964-10G A126964-1G A126964-250MG A126964-25G A126964-5G
77108-936EA 172.01 USD
77108-936 77108-938 77108-940 77108-942 77108-944 77108-946
Boc-S-trityl-D-cysteine 97%
Boc-S-trityl-D-cysteine

Precaución: for Research Use Only

Formula: C₂₇H₂₉NO₄S
MDL Number: MFCD00236839
CAS Number: 87494-13-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR