4-Nitro-2-picoline-N-oxide 98%

Supplier: Ambeed
Danger

Synonyms: 2-Methyl-4-nitropyridine-1-oxide, 4-Nitro-2-picoline N-oxide, 2-Methyl-4-nitropyridine-N-oxide

A126845-10G A126845-1G A126845-25G A126845-5G
77108-922EA 15.92 USD
77108-922 77108-924 77108-926 77108-928
4-Nitro-2-picoline-N-oxide 98%
4-Nitro-2-picoline-N-oxide

Precaución: for Research Use Only

Formula: C₆H₆N₂O₃
MW: 154.13 g/mol
MDL Number: MFCD00014625
CAS Number: 5470-66-6
UN: 2811
ADR: 6.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR