(S)-2-(((Benzyloxy)carbonyl)amino)-3-(4-cyanophenyl)propanoic acid 95%

Supplier: Ambeed
Warning

A126766-100MG A126766-250MG
77108-916EA 15.44 USD
77108-916 77108-918
(S)-2-(((Benzyloxy)carbonyl)amino)-3-(4-cyanophenyl)propanoic acid 95%
(S)-2-(((Benzyloxy)carbonyl)amino)-3-(4-cyanophenyl)propanoic acid

Precaución: for Research Use Only

Formula: C₁₈H₁₆N₂O₄
MDL Number: MFCD02094456
CAS Number: 131669-07-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR